Product Details
- SNP ID
-
rs192070793
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:47280327 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAAGGCACAGAAAATCACACTGTT[C/T]ACGGGGTAATGCCTCCGGAAGAAGA
- Phenotype
-
MIM: 606825
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TNS3
PubMed Links
Gene Details
- Gene
- TNS3
- Gene Name
- tensin 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022748.11 |
4043 |
Silent Mutation |
GTA,GTG |
V1375V |
NP_073585.8 |
XM_011515476.2 |
4043 |
Silent Mutation |
GTA,GTG |
V1491V |
XP_011513778.1 |
XM_011515477.2 |
4043 |
Silent Mutation |
GTA,GTG |
V1478V |
XP_011513779.1 |
XM_011515478.2 |
4043 |
Silent Mutation |
GTA,GTG |
V1464V |
XP_011513780.1 |
XM_011515479.2 |
4043 |
Silent Mutation |
GTA,GTG |
V1418V |
XP_011513781.1 |
XM_011515480.2 |
4043 |
Silent Mutation |
GTA,GTG |
V1375V |
XP_011513782.1 |
XM_011515481.2 |
4043 |
Silent Mutation |
GTA,GTG |
V1375V |
XP_011513783.1 |
XM_011515482.2 |
4043 |
Silent Mutation |
GTA,GTG |
V1375V |
XP_011513784.1 |
XM_011515483.2 |
4043 |
Silent Mutation |
GTA,GTG |
V1375V |
XP_011513785.1 |
XM_011515484.2 |
4043 |
Silent Mutation |
GTA,GTG |
V932V |
XP_011513786.1 |
XM_017012537.1 |
4043 |
Silent Mutation |
GTA,GTG |
V1478V |
XP_016868026.1 |
XM_017012538.1 |
4043 |
Silent Mutation |
GTA,GTG |
V1489V |
XP_016868027.1 |
XM_017012539.1 |
4043 |
Silent Mutation |
GTA,GTG |
V1375V |
XP_016868028.1 |
View Full Product Details