Product Details

SNP ID
rs191963770
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:34987237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGAAAAGGCAGCATCACATAAA[C/T]AAACAGTTGAAAAGGGTAATAAAAA
Phenotype
MIM: 613567
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC653160 PubMed Links

Gene Details

Gene
LOC653160
Gene Name
uncharacterized LOC653160
There are no transcripts associated with this gene.

Gene
TMEM35B
Gene Name
transmembrane protein 35B
There are no transcripts associated with this gene.

Gene
ZMYM6
Gene Name
zinc finger MYM-type containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007167.3 4073 Missense Mutation TAT,TGT Y1282C NP_009098.3

View Full Product Details