Product Details

SNP ID
rs192189898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64691509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGATCTTCGACAGTGGGTGAGG[A/G]GCATCAGGAGCACCACAGAGGCCCT
Phenotype
MIM: 604152
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OAZ2 PubMed Links

Gene Details

Gene
OAZ2
Gene Name
ornithine decarboxylase antizyme 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301302.1 336 UTR 3 NP_001288231.1
NM_002537.3 336 UTR 3 NP_002528.1
Gene
ZNF609
Gene Name
zinc finger protein 609
There are no transcripts associated with this gene.

View Full Product Details