Product Details

SNP ID
rs193233043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30385143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGCTTATGCTTCAAACCACCCC[A/G]TAATGAAAACTGGTGAGCCTCATTG
Phenotype
MIM: 603102
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPD PubMed Links

Gene Details

Gene
CPD
Gene Name
carboxypeptidase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199775.1 320 Missense Mutation ATA,GTA I54V NP_001186704.1
NM_001304.4 320 Missense Mutation ATA,GTA I301V NP_001295.2

View Full Product Details