Product Details

SNP ID
rs193031994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45818070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCAGCATGGGCAGGGGGCTCTG[A/C]TCCATCAGCTGCTGCATCACCACGG
Phenotype
MIM: 607548 MIM: 602388
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RSPH6A PubMed Links

Gene Details

Gene
RSPH6A
Gene Name
radial spoke head 6 homolog A
There are no transcripts associated with this gene.

Gene
SYMPK
Gene Name
symplekin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004819.2 3168 Missense Mutation GAG,GAT E990D NP_004810.2
XM_005259286.1 3168 Missense Mutation GAG,GAT E990D XP_005259343.1
XM_011527354.1 3168 Missense Mutation GAG,GAT E990D XP_011525656.1

View Full Product Details