Product Details

SNP ID
rs192733942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:201129757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCCACTGGAAAGGATTCTGAAA[A/G]AAATGAAGTCAGCCCTCAGAAATGA
Phenotype
MIM: 603599
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFLAR PubMed Links

Gene Details

Gene
CFLAR
Gene Name
CASP8 and FADD like apoptosis regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127183.2 300 UTR 5 NP_001120655.1
NM_001127184.2 300 UTR 5 NP_001120656.1
NM_001202515.1 300 Intron NP_001189444.1
NM_001202516.1 300 UTR 5 NP_001189445.1
NM_001202517.1 300 Intron NP_001189446.1
NM_001202518.1 300 Intron NP_001189447.1
NM_001202519.1 300 Intron NP_001189448.1
NM_001308042.1 300 UTR 5 NP_001294971.1
NM_001308043.1 300 UTR 5 NP_001294972.1
NM_003879.5 300 UTR 5 NP_003870.4
XM_017005189.1 300 UTR 5 XP_016860678.1
XM_017005190.1 300 UTR 5 XP_016860679.1
XM_017005191.1 300 UTR 5 XP_016860680.1
XM_017005192.1 300 UTR 5 XP_016860681.1
XM_017005193.1 300 UTR 5 XP_016860682.1
XM_017005194.1 300 UTR 5 XP_016860683.1
XM_017005195.1 300 UTR 5 XP_016860684.1
XM_017005196.1 300 Intron XP_016860685.1
XM_017005197.1 300 UTR 5 XP_016860686.1
XM_017005198.1 300 UTR 5 XP_016860687.1

View Full Product Details