Product Details

SNP ID
rs27961
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:66147524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTGACCCCCTTCAAGTAACTAAA[A/C]CCTGAAGGGTAAGCCTACCCTGATT
Phenotype
MIM: 609268
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SREK1 PubMed Links

Gene Details

Gene
SREK1
Gene Name
splicing regulatory glutamic acid and lysine rich protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077199.2 Intron NP_001070667.1
NM_001270492.1 Intron NP_001257421.1
NM_001323527.1 Intron NP_001310456.1
NM_001323529.1 Intron NP_001310458.1
NM_001323533.1 Intron NP_001310462.1
NM_001323534.1 Intron NP_001310463.1
NM_001323535.1 Intron NP_001310464.1
NM_139168.3 Intron NP_631907.1
XM_011543171.2 Intron XP_011541473.1
XM_017009055.1 Intron XP_016864544.1
XM_017009056.1 Intron XP_016864545.1

View Full Product Details