Product Details

SNP ID
rs6671285
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:89107825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATACAGTAAGGGTGGTGCATAT[A/G]AGAGAAAAAAATCTACCCAGAACAG
Phenotype
MIM: 600412
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GBP2 PubMed Links

Gene Details

Gene
GBP2
Gene Name
guanylate binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004120.4 2440 UTR 3 NP_004111.2

View Full Product Details