Product Details

SNP ID
rs6754850
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:64528381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATGAGCAAACAAGAAAAAAAAT[A/C]TGCCCTCCTACAGCTTCAGTTTCTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AFTPH PubMed Links

Gene Details

Gene
AFTPH
Gene Name
aftiphilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002243.2 Intron NP_001002243.1
NM_017657.4 Intron NP_060127.3
NM_203437.3 Intron NP_982261.2
XM_005264380.1 Intron XP_005264437.1
Gene
LOC101927402
Gene Name
uncharacterized LOC101927402
There are no transcripts associated with this gene.

Gene
MIR4434
Gene Name
microRNA 4434
There are no transcripts associated with this gene.

View Full Product Details