Product Details

SNP ID
rs719349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52068338 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTATTTGGAAAAAATATTTCCAAT[G/A]TATAGCACCGTTATTTCTCTCCATT
Phenotype
MIM: 616731
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
LOC107984562 PubMed Links

Gene Details

Gene
LOC107984562
Gene Name
serine/threonine-protein kinase Nek5-like
There are no transcripts associated with this gene.

Gene
NEK5
Gene Name
NIMA related kinase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199289.2 Intron NP_954983.1
XM_006719808.3 Intron XP_006719871.2
XM_011535066.2 Intron XP_011533368.1
XM_011535068.2 Intron XP_011533370.1
XM_017020555.1 Intron XP_016876044.1
XM_017020556.1 Intron XP_016876045.1
XM_017020557.1 Intron XP_016876046.1

View Full Product Details