Product Details
- SNP ID
-
rs7327634
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:52013609 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTCCCACCTAAATTATTTGAAGGA[A/C]TTGCCAGAAAAACCTTTACATTATA
- Phenotype
-
MIM: 613666
MIM: 606882
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ALG11
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7328090] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALG11
- Gene Name
- ALG11, alpha-1,2-mannosyltransferase
- Gene
- ATP7B
- Gene Name
- ATPase copper transporting beta
There are no transcripts associated with this gene.
View Full Product Details