Product Details
- SNP ID
-
rs7530165
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
45
- Location
-
Chr.1:22433832 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTAATTATTTAATTAAACTGCATA[A/G]TATGCCATTATATGGATGTATCACA
- Phenotype
-
MIM: 612106
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZBTB40
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7532596] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZBTB40
- Gene Name
- zinc finger and BTB domain containing 40
View Full Product Details