Product Details

SNP ID
rs7519323
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:89109405 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAACTAATAGTTCCTCTACCATTA[A/G]AGGAAAATTTGATCACATAATGTAA
Phenotype
MIM: 600412
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GBP2 PubMed Links

Gene Details

Gene
GBP2
Gene Name
guanylate binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004120.4 Intron NP_004111.2

View Full Product Details