Product Details

SNP ID
rs773904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16889320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGTGAGTGGTCCTGGCTTTGGG[A/G]TGCAGAGATGGGGTTACATCAGAGA
Phenotype
MIM: 608841 MIM: 602779 MIM: 607777
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPAMD8 PubMed Links

Gene Details

Gene
CPAMD8
Gene Name
C3 and PZP like, alpha-2-macroglobulin domain containing 8
There are no transcripts associated with this gene.

Gene
F2RL3
Gene Name
F2R like thrombin/trypsin receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003950.2 Intron NP_003941.2
XM_005260139.2 Intron XP_005260196.1
Gene
SIN3B
Gene Name
SIN3 transcription regulator family member B
There are no transcripts associated with this gene.

View Full Product Details