Product Details
- SNP ID
-
rs7883011
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:16808161 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGATTGCGAAATATATCCTTTGAC[C/T]GGAGGGGAGTGGCTAGGGTGAGGGA
- Phenotype
-
MIM: 300677
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TXLNG
PubMed Links
Gene Details
- Gene
- TXLNG
- Gene Name
- taxilin gamma
View Full Product Details