Product Details

SNP ID
rs201167753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:13283759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAAGGATGGAAGAAAACAGTGTC[A/G]CCCTTCTCCATCACCAGGTGCACCC
Phenotype
MIM: 602026
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHYH PubMed Links

Gene Details

Gene
PHYH
Gene Name
phytanoyl-CoA 2-hydroxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037537.1 853 Silent Mutation GGC,GGT G153G NP_001032626.1
NM_001323080.1 853 Silent Mutation GGC,GGT G153G NP_001310009.1
NM_001323082.1 853 Silent Mutation GGC,GGT G255G NP_001310011.1
NM_001323083.1 853 Silent Mutation GGC,GGT G165G NP_001310012.1
NM_001323084.1 853 Silent Mutation GGC,GGT G155G NP_001310013.1
NM_006214.3 853 Silent Mutation GGC,GGT G253G NP_006205.1

View Full Product Details