Product Details

SNP ID
rs199905401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:26217963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCTGCAGCTGCTCCAAAGTGGA[C/T]GTCAACTACGCGTTTCTCCATGCAA
Phenotype
MIM: 138275 MIM: 606808
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAD2 PubMed Links

Gene Details

Gene
GAD2
Gene Name
glutamate decarboxylase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000818.2 761 Silent Mutation GAC,GAT D86D NP_000809.1
NM_001134366.1 761 Silent Mutation GAC,GAT D86D NP_001127838.1
Gene
MYO3A
Gene Name
myosin IIIA
There are no transcripts associated with this gene.

View Full Product Details