Product Details

SNP ID
rs201906236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:110207891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCCGCGCCCCCGGCTGTGCCCC[C/G]CGCCGTGGCCGCGCCCCAGCCCCCG
Phenotype
MIM: 600020
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MXI1 PubMed Links

Gene Details

Gene
MXI1
Gene Name
MAX interactor 1, dimerization protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008541.1 287 Intron NP_001008541.1
NM_005962.4 287 Intron NP_005953.4
NM_130439.3 287 Missense Mutation CCC,CGC P28R NP_569157.2

View Full Product Details