Product Details

SNP ID
rs201017802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3090614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAGCTCTGGGCAGGACTCAGGC[A/G]TGGATCCGCTGCTCTCCGTGGCCTG
Phenotype
MIM: 606733
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OSBPL5 PubMed Links

Gene Details

Gene
OSBPL5
Gene Name
oxysterol binding protein like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144063.1 2847 Missense Mutation ACG,ATG T713M NP_001137535.1
NM_020896.3 2847 Missense Mutation ACG,ATG T781M NP_065947.1
NM_145638.2 2847 Missense Mutation ACG,ATG T713M NP_663613.1
XM_011519873.2 2847 Missense Mutation ACG,ATG T781M XP_011518175.1
XM_017017163.1 2847 Missense Mutation ACG,ATG T781M XP_016872652.1
XM_017017164.1 2847 Missense Mutation ACG,ATG T411M XP_016872653.1

View Full Product Details