Product Details

SNP ID
rs200803638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56543088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTAGTGAGGAAGAAGTACATGGGC[A/G]TGTGAAGGCGAGAGTCCAGTCTCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5M11 PubMed Links

Gene Details

Gene
OR5M11
Gene Name
olfactory receptor family 5 subfamily M member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005245.1 170 Missense Mutation ACG,ATG T57M NP_001005245.1

View Full Product Details