Product Details

SNP ID
rs200875557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47179487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAAGAAAAGAAGCCGCTGGCGCT[C/T]GGCGGCCTGGCAGGTCTCAAAGGCC
Phenotype
MIM: 606908 MIM: 606513
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARFGAP2 PubMed Links

Gene Details

Gene
ARFGAP2
Gene Name
ADP ribosylation factor GTPase activating protein 2
There are no transcripts associated with this gene.

Gene
MIR6745
Gene Name
microRNA 6745
There are no transcripts associated with this gene.

Gene
PACSIN3
Gene Name
protein kinase C and casein kinase substrate in neurons 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184974.1 917 Missense Mutation AAG,GAG K235E NP_001171903.1
NM_001184975.1 917 Missense Mutation AAG,GAG K235E NP_001171904.1
NM_016223.4 917 Missense Mutation AAG,GAG K235E NP_057307.2

View Full Product Details