Product Details

SNP ID
rs201036032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6891804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTAGAAAATGCCTTGAGACTCC[A/C]CACAGTTGACTTCATCTTGACCACA
Phenotype
MIM: 608494
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR2D2 PubMed Links

Gene Details

Gene
OR2D2
Gene Name
olfactory receptor family 2 subfamily D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003700.1 697 Missense Mutation NP_003691.1

View Full Product Details