Product Details

SNP ID
rs200118587
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:114063353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTGCAGAACCCTAGCCACCCCA[C/T]GGGGCTACTGTGCAAGGCCAACCAG
Phenotype
MIM: 176797
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZBTB16 PubMed Links

Gene Details

Gene
ZBTB16
Gene Name
zinc finger and BTB domain containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018011.1 757 Missense Mutation ACG,ATG T18M NP_001018011.1
NM_006006.4 757 Missense Mutation ACG,ATG T18M NP_005997.2
XM_005271658.4 757 Missense Mutation ACG,ATG T18M XP_005271715.1
XM_017018257.1 757 Missense Mutation ACG,ATG T18M XP_016873746.1
XM_017018258.1 757 Missense Mutation ACG,ATG T18M XP_016873747.1
XM_017018259.1 757 Missense Mutation ACG,ATG T18M XP_016873748.1

View Full Product Details