Product Details

SNP ID
rs200033070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:101453084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCATAGCGGAGACTTGAGATGTC[C/T]TGCTTAATTTCCTTCAGTTCCCCTT
Phenotype
MIM: 603652
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRPC6 PubMed Links

Gene Details

Gene
TRPC6
Gene Name
transient receptor potential cation channel subfamily C member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004621.5 2594 Silent Mutation CAA,CAG Q889Q NP_004612.2
XM_011542968.2 2594 Silent Mutation CAA,CAG Q834Q XP_011541270.1
XM_017018221.1 2594 Silent Mutation CAA,CAG Q773Q XP_016873710.1
XM_017018222.1 2594 Silent Mutation CAA,CAG Q803Q XP_016873711.1

View Full Product Details