Product Details

SNP ID
rs201591455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58422378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACACTTTCTTGAATGCATTCTGG[A/T]CTTCTCTGTTCCTCAGGCTGTAGAC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR5B2 PubMed Links

Gene Details

Gene
OR5B2
Gene Name
olfactory receptor family 5 subfamily B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005566.2 936 Missense Mutation GAC,GTC D295V NP_001005566.1

View Full Product Details