Product Details

SNP ID
rs201719402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:16744664 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGAGAGAAAACAAAAGTTCTTG[A/C]GACTTATGGGTGCAGGAAAGGTAAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C11orf58 PubMed Links

Gene Details

Gene
C11orf58
Gene Name
chromosome 11 open reading frame 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014267.5 2564 Silent Mutation AGA,CGA R43R NP_055082.1
XM_017017143.1 2564 UTR 5 XP_016872632.1

View Full Product Details