Product Details

SNP ID
rs201770114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:27338896 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGAATTACCAGAGATGATGACTT[G/T]TGTGGCTGACTTATCACAGGTCTTT
Phenotype
MIM: 612324
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC34 PubMed Links

Gene Details

Gene
CCDC34
Gene Name
coiled-coil domain containing 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030771.1 1101 Missense Mutation CAA,CAC Q349H NP_110398.1
NM_080654.2 1101 Intron NP_542385.1

View Full Product Details