Product Details

SNP ID
rs201723938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56542581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCCTTCCCTCTGCTGATTTGATC[C/T]GGAGGATGGCAGCAAGAATGAAGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5M11 PubMed Links

Gene Details

Gene
OR5M11
Gene Name
olfactory receptor family 5 subfamily M member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005245.1 677 Missense Mutation CAG,CGG Q226R NP_001005245.1

View Full Product Details