Product Details

SNP ID
rs201877831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124029894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTGTTCCCAAGTGGCAGGGCTA[C/T]CTCCTTCCACAGCTGCATGGCTCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10G7 PubMed Links

Gene Details

Gene
OR10G7
Gene Name
olfactory receptor family 10 subfamily G member 7
There are no transcripts associated with this gene.

Gene
OR10G8
Gene Name
olfactory receptor family 10 subfamily G member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004464.1 Intron NP_001004464.1
Gene
OR10G9
Gene Name
olfactory receptor family 10 subfamily G member 9
There are no transcripts associated with this gene.

View Full Product Details