Product Details

SNP ID
rs201831239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65713352 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTGGACCTATCTCTACAGCCGG[C/T]CTCGGCGCAGGCCAGCGGGAAGACC
Phenotype
MIM: 601409 MIM: 610330
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KAT5 PubMed Links

Gene Details

Gene
KAT5
Gene Name
lysine acetyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206833.1 556 Intron NP_001193762.1
NM_006388.3 556 Missense Mutation GCC,GTC A97V NP_006379.2
NM_182709.2 556 Intron NP_874368.1
NM_182710.2 556 Missense Mutation GCC,GTC A130V NP_874369.1
XM_006718421.2 556 Missense Mutation GCC,GTC A106V XP_006718484.1
Gene
RNASEH2C
Gene Name
ribonuclease H2 subunit C
There are no transcripts associated with this gene.

View Full Product Details