Product Details

SNP ID
rs202043963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56030669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATCACTCCTAAAATGCTGGCAA[A/G]CTTCTTGGCATCCAGGAAAAGCATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5AS1 PubMed Links

Gene Details

Gene
OR5AS1
Gene Name
olfactory receptor family 5 subfamily AS member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001921.1 251 Missense Mutation AAC,AGC N84S NP_001001921.1

View Full Product Details