Product Details

SNP ID
rs202091355
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:101455082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATCTTCTGAGCTCCTTATACTT[G/T]GTTGTTTATTGTGCCCAACCTGTAA
Phenotype
MIM: 603652
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TRPC6 PubMed Links

Gene Details

Gene
TRPC6
Gene Name
transient receptor potential cation channel subfamily C member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004621.5 2431 Missense Mutation CAA,CCA Q835P NP_004612.2
XM_011542968.2 2431 Missense Mutation CAA,CCA Q780P XP_011541270.1
XM_017018221.1 2431 Missense Mutation CAA,CCA Q719P XP_016873710.1
XM_017018222.1 2431 Missense Mutation CAA,CCA Q749P XP_016873711.1

View Full Product Details