Product Details

SNP ID
rs201813599
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124249784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTGTTCTTAGGAATCTATGTG[A/G]TCACAGTGGTGGGCAACCTGGGCAT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR8G1 PubMed Links
Additional Information
For this assay, SNP(s) [rs138496920] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
OR8G1
Gene Name
olfactory receptor family 8 subfamily G member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002905.1 109 Missense Mutation ATC,GTC I37V NP_001002905.1
Gene
OR8G5
Gene Name
olfactory receptor family 8 subfamily G member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005198.1 109 Intron NP_001005198.1
XM_011542662.2 109 Intron XP_011540964.1
XM_011542663.2 109 Intron XP_011540965.1
XM_011542664.2 109 Intron XP_011540966.1
XM_011542665.2 109 Intron XP_011540967.1
XM_011542666.2 109 Intron XP_011540968.1
XM_011542667.2 109 Intron XP_011540969.1

View Full Product Details