Product Details

SNP ID
rs202118284
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:32389064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCCCGAGGGAGACCCCTCAAAG[C/T]GCCAGCTGGAGTTTGGTCATGTTTC
Phenotype
MIM: 607102
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WT1 PubMed Links

Gene Details

Gene
WT1
Gene Name
Wilms tumor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000378.4 1139 Silent Mutation GCA,GCG A496A NP_000369.3
NM_001198551.1 1139 Silent Mutation GCA,GCG A301A NP_001185480.1
NM_001198552.1 1139 Silent Mutation GCA,GCG A287A NP_001185481.1
NM_024424.3 1139 Silent Mutation GCA,GCG A513A NP_077742.2
NM_024426.4 1139 Silent Mutation GCA,GCG A516A NP_077744.3

View Full Product Details