Product Details

SNP ID
rs201106689
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6891612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTGGCTACTTTCCTCAGAGCTG[A/C]CTTCACATCCTTGTTTCTCAGGCTA
Phenotype
MIM: 608494
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR2D2 PubMed Links

Gene Details

Gene
OR2D2
Gene Name
olfactory receptor family 2 subfamily D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003700.1 889 Missense Mutation GCA,TCA A297S NP_003691.1

View Full Product Details