Product Details

SNP ID
rs201165872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:613076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAAGCCCAGGTAGATGGTATAGC[A/G]TGGGGAGCCACGGCGCTGCCGTGCC
Phenotype
MIM: 606839 MIM: 605047 MIM: 611780
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDHR5 PubMed Links

Gene Details

Gene
CDHR5
Gene Name
cadherin related family member 5
There are no transcripts associated with this gene.

Gene
IRF7
Gene Name
interferon regulatory factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001572.3 1728 Missense Mutation CGC,TGC R427C NP_001563.2
NM_004029.2 1728 Missense Mutation CGC,TGC R398C NP_004020.1
NM_004031.2 1728 Missense Mutation CGC,TGC R440C NP_004022.2
XM_005252906.3 1728 Missense Mutation CGC,TGC R440C XP_005252963.1
XM_005252907.3 1728 Missense Mutation CGC,TGC R439C XP_005252964.1
XM_005252909.3 1728 Missense Mutation CGC,TGC R411C XP_005252966.1
XM_011520066.2 1728 Missense Mutation CGC,TGC R426C XP_011518368.1
XM_017017674.1 1728 Missense Mutation CGC,TGC R134C XP_016873163.1
Gene
PHRF1
Gene Name
PHD and ring finger domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286581.1 1728 Intron NP_001273510.1
NM_001286582.1 1728 Intron NP_001273511.1
NM_001286583.1 1728 Intron NP_001273512.1
NM_020901.3 1728 Intron NP_065952.2
XM_005253025.4 1728 Intron XP_005253082.1
XM_005253027.3 1728 Intron XP_005253084.1
XM_011520236.2 1728 Intron XP_011518538.1
XM_011520237.2 1728 Intron XP_011518539.1

View Full Product Details