Product Details
- SNP ID
-
rs199613662
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52491555 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGTCCTCCACCAGGTCCTGCATG[C/T]CTCTGAGCTCTGAGTCCAGGCGGCC
- Phenotype
-
MIM: 148041
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KRT6A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201663666,rs534124939] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KRT6A
- Gene Name
- keratin 6A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005554.3 |
931 |
Missense Mutation |
GAC,GGC |
D241G |
NP_005545.1 |
View Full Product Details