Product Details

SNP ID
rs199512241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:130872874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATGGTGGTACTGGAAAAACGAC[C/T]TTCGTGAAACGTCATTTGACTGGTG
Phenotype
MIM: 601179
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAN PubMed Links

Gene Details

Gene
RAN
Gene Name
RAN, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300796.1 172 UTR 5 NP_001287725.1
NM_001300797.1 172 UTR 5 NP_001287726.1
NM_006325.4 172 Silent Mutation ACC,ACT T25T NP_006316.1
XM_017019772.1 172 Silent Mutation ACC,ACT T25T XP_016875261.1
XM_017019773.1 172 UTR 5 XP_016875262.1

View Full Product Details