Product Details
- SNP ID
-
rs200885137
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:25108608 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGGCACACAGAGTTGACAAACTG[A/T]CAGTTGGAACTCCTGACTTTCTCCA
- Phenotype
-
MIM: 616906
MIM: 602003
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CASC1
PubMed Links
Gene Details
- Gene
- CASC1
- Gene Name
- cancer susceptibility candidate 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001082972.2 |
2272 |
UTR 3 |
|
|
NP_001076441.1 |
NM_001082973.2 |
2272 |
Nonsense Mutation |
TGA,TGT |
*697C |
NP_001076442.1 |
NM_001204101.2 |
2272 |
Nonsense Mutation |
TGA,TGT |
*638C |
NP_001191030.1 |
NM_001204102.2 |
2272 |
Nonsense Mutation |
TGA,TGT |
*657C |
NP_001191031.1 |
NM_001319977.1 |
2272 |
UTR 3 |
|
|
NP_001306906.1 |
NM_001319978.1 |
2272 |
Nonsense Mutation |
TGA,TGT |
*657C |
NP_001306907.1 |
NM_018272.4 |
2272 |
Nonsense Mutation |
TGA,TGT |
*703C |
NP_060742.3 |
XM_005253406.1 |
2272 |
Nonsense Mutation |
TGA,TGT |
*761C |
XP_005253463.1 |
XM_005253408.1 |
2272 |
Nonsense Mutation |
TGA,TGT |
*585C |
XP_005253465.1 |
XM_005253409.1 |
2272 |
Nonsense Mutation |
TGA,TGT |
*585C |
XP_005253466.1 |
XM_006719101.1 |
2272 |
Nonsense Mutation |
TGA,TGT |
*655C |
XP_006719164.1 |
XM_011520723.1 |
2272 |
Nonsense Mutation |
TGA,TGT |
*761C |
XP_011519025.1 |
XM_011520726.1 |
2272 |
Intron |
|
|
XP_011519028.1 |
XM_011520727.1 |
2272 |
Intron |
|
|
XP_011519029.1 |
XM_017019560.1 |
2272 |
Nonsense Mutation |
TGA,TGT |
*755C |
XP_016875049.1 |
XM_017019561.1 |
2272 |
Nonsense Mutation |
TGA,TGT |
*657C |
XP_016875050.1 |
XM_017019562.1 |
2272 |
UTR 3 |
|
|
XP_016875051.1 |
XM_017019563.1 |
2272 |
Nonsense Mutation |
TGA,TGT |
*597C |
XP_016875052.1 |
XM_017019564.1 |
2272 |
Nonsense Mutation |
TGA,TGT |
*591C |
XP_016875053.1 |
XM_017019565.1 |
2272 |
UTR 3 |
|
|
XP_016875054.1 |
XM_017019566.1 |
2272 |
Nonsense Mutation |
TGA,TGT |
*551C |
XP_016875055.1 |
XM_017019567.1 |
2272 |
Nonsense Mutation |
TGA,TGT |
*532C |
XP_016875056.1 |
XM_017019568.1 |
2272 |
UTR 3 |
|
|
XP_016875057.1 |
- Gene
- LRMP
- Gene Name
- lymphoid restricted membrane protein
View Full Product Details