Product Details

SNP ID
rs200166993
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54325172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGCTGCGGGGCAAGATGGAGGC[A/G]CTGATTTTGGTAGGAGCTGGAGGGG
Phenotype
MIM: 615472
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPZ1 PubMed Links

Gene Details

Gene
COPZ1
Gene Name
coatomer protein complex subunit zeta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271734.1 64 Silent Mutation GCA,GCG A3A NP_001258663.1
NM_001271735.1 64 Silent Mutation GCA,GCG A3A NP_001258664.1
NM_001271736.1 64 UTR 5 NP_001258665.1
NM_016057.2 64 Silent Mutation GCA,GCG A3A NP_057141.1
XM_017018997.1 64 Silent Mutation GCA,GCG A3A XP_016874486.1

View Full Product Details