Product Details

SNP ID
rs200767486
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:118139504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAGGGCAATGACATCAGCAGTG[A/G]CACAGTCCTCTCCGATTATGTGGGC
Phenotype
MIM: 604591
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PEBP1 PubMed Links

Gene Details

Gene
PEBP1
Gene Name
phosphatidylethanolamine binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002567.3 625 Missense Mutation GAC,GGC D100G NP_002558.1

View Full Product Details