Product Details

SNP ID
rs199791754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49348902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGGGGCTCCTGGTGACCTATGC[C/T]CTCTGGGCTGTGGGGGGCCCTGCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC22 PubMed Links

Gene Details

Gene
DNAJC22
Gene Name
DnaJ heat shock protein family (Hsp40) member C22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304944.1 772 Silent Mutation GCC,GCT A10A NP_001291873.1
NM_024902.3 772 Silent Mutation GCC,GCT A10A NP_079178.2
XM_005269155.2 772 Silent Mutation GCC,GCT A10A XP_005269212.1
XM_005269156.2 772 Silent Mutation GCC,GCT A10A XP_005269213.1
XM_005269157.2 772 Silent Mutation GCC,GCT A10A XP_005269214.1

View Full Product Details