Product Details
- SNP ID
-
rs202197926
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52515095 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACACTGAGCCCACCACCTAGGCC[A/G]ACACCCCCACTGCTGCTGGAGTAGT
- Phenotype
-
MIM: 148040
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KRT5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11549949] are located under a probe and SNP(s) [rs11549950] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KRT5
- Gene Name
- keratin 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000424.3 |
1783 |
Silent Mutation |
GTC,GTT |
V540V |
NP_000415.2 |
View Full Product Details