Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286233.1 | 1035 | Missense Mutation | NP_001273162.1 | ||
NM_001286234.1 | 1035 | Missense Mutation | NP_001273163.1 | ||
NM_001286235.1 | 1035 | Missense Mutation | NP_001273164.1 | ||
NM_001286236.1 | 1035 | Missense Mutation | NP_001273165.1 | ||
NM_001286237.1 | 1035 | Intron | NP_001273166.1 | ||
NM_153449.3 | 1035 | Missense Mutation | NP_703150.1 | ||
XM_005253315.3 | 1035 | Intron | XP_005253372.1 | ||
XM_005253317.4 | 1035 | Missense Mutation | XP_005253374.1 | ||
XM_011520562.1 | 1035 | Missense Mutation | XP_011518864.1 | ||
XM_011520563.2 | 1035 | Intron | XP_011518865.1 | ||
XM_011520564.2 | 1035 | Missense Mutation | XP_011518866.1 | ||
XM_011520565.2 | 1035 | Missense Mutation | XP_011518867.1 | ||
XM_017018841.1 | 1035 | Intron | XP_016874330.1 | ||
XM_017018842.1 | 1035 | Intron | XP_016874331.1 | ||
XM_017018843.1 | 1035 | Missense Mutation | XP_016874332.1 | ||
XM_017018844.1 | 1035 | Intron | XP_016874333.1 | ||
XM_017018845.1 | 1035 | Missense Mutation | XP_016874334.1 | ||
XM_017018846.1 | 1035 | Intron | XP_016874335.1 | ||
XM_017018847.1 | 1035 | Intron | XP_016874336.1 |