Product Details

SNP ID
rs201244234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49348882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTTGCCCTAGAGGATGGCCAAG[G/T]GGCTCCTGGTGACCTATGCCCTCTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC22 PubMed Links

Gene Details

Gene
DNAJC22
Gene Name
DnaJ heat shock protein family (Hsp40) member C22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304944.1 752 Missense Mutation GGG,TGG G4W NP_001291873.1
NM_024902.3 752 Missense Mutation GGG,TGG G4W NP_079178.2
XM_005269155.2 752 Missense Mutation GGG,TGG G4W XP_005269212.1
XM_005269156.2 752 Missense Mutation GGG,TGG G4W XP_005269213.1
XM_005269157.2 752 Missense Mutation GGG,TGG G4W XP_005269214.1

View Full Product Details