Product Details
- SNP ID
-
hCV189963919
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:24426473 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACTATAGTTAAAGCCACCTGGAAA[T/A]AGAAGCGTCATCCATTTTCATCAGT
- Phenotype
-
MIM: 607519
- Polymorphism
- T/A, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PARP4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7571] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PARP4
- Gene Name
- poly(ADP-ribose) polymerase family member 4
There are no transcripts associated with this gene.
View Full Product Details