Product Details

SNP ID
rs202162563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52844928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTACACTCAGCGGTGCACGCCC[A/G]CAGTCCTAATACGAAAGGGAAAAGG
Phenotype
MIM: 603580
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDH8 PubMed Links

Gene Details

Gene
PCDH8
Gene Name
protocadherin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002590.3 3049 Missense Mutation CGG,TGG R949W NP_002581.2
NM_032949.2 3049 Missense Mutation CGG,TGG R852W NP_116567.1

View Full Product Details