Product Details

SNP ID
rs201957260
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23033370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTGGTTGCAGCTTAACTCACCA[A/G]GGCCTCTCTTATCCCAGCCACAGAT
Phenotype
MIM: 611137 MIM: 600306
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSMB11 PubMed Links

Gene Details

Gene
PSMB11
Gene Name
proteasome subunit beta 11
There are no transcripts associated with this gene.

Gene
PSMB5
Gene Name
proteasome subunit beta 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130725.1 762 Missense Mutation CCT,CTT P65L NP_001124197.1
NM_001144932.2 762 Missense Mutation CCT,CTT P168L NP_001138404.1
NM_002797.4 762 Missense Mutation CCT,CTT P168L NP_002788.1
XM_005267871.2 762 Missense Mutation CCT,CTT P168L XP_005267928.1

View Full Product Details