Product Details

SNP ID
rs201850675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24631926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATGGTACTGTCGTAATAATGGC[A/G]TAAGTCAGATTCGCACTTTCGATCT
Phenotype
MIM: 123910
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GZMB PubMed Links

Gene Details

Gene
GZMB
Gene Name
granzyme B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004131.4 582 Missense Mutation CGC,TGC R178C NP_004122.2
XM_011536685.2 582 Missense Mutation CGC,TGC R166C XP_011534987.1

View Full Product Details