Product Details

SNP ID
rs199941584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24631915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACGCACAACTCAATGGTACTGTC[A/G]TAATAATGGCGTAAGTCAGATTCGC
Phenotype
MIM: 123910
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GZMB PubMed Links

Gene Details

Gene
GZMB
Gene Name
granzyme B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004131.4 593 Silent Mutation TAC,TAT Y181Y NP_004122.2
XM_011536685.2 593 Silent Mutation TAC,TAT Y169Y XP_011534987.1

View Full Product Details